Order Entry
China
ContactUsLinkComponent
Hexane (mixture of isomers), extra dry 99.9%, AcroSeal®
 
 
 
 
Hexane (mixture of isomers), extra dry 99.9%, AcroSeal®
  ACRO610371000
 :  
undefined
 
 
 
 
Hexane (mixture of isomers), extra dry 99.9%, AcroSeal®
  ACRO610371000
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    Colorless Liquid
  • Assay:
    ≥99.9 % total C6 isomers
  • Water:
    ≤50 ppm

 

 

 :
 :86.18 g/mol
 :13,04712
 :69 °C (1013 hPa)
 :–95 °C
 :0.659 g/cm³
 :–22 °C
 :Ambient
 :203-777-6
 :1208
 :3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
 :92112-69-1
 :H₃C(CH₂)₄CH₃
 :MFCD02179311