1,2,6-Hexanetriol 97+%, extra pure
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1,2,6-Trihydroxyhexane
- CAS Number:106-69-4
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00002976
- Molecular Weight:134.18 g/mol
- Boiling Pt:178 °C (6.7 mmHg)
- Melting Pt:25…32 °C
- Density:1.109 g/cm³
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:203-424-6
2 Options Available Below
Product Details & Documents
Synonyms: 1,2,6-Trihydroxyhexane
- CAS Number:106-69-4
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00002976
- Molecular Weight:134.18 g/mol
- Boiling Pt:178 °C (6.7 mmHg)
- Melting Pt:25…32 °C
- Density:1.109 g/cm³
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:203-424-6
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid (May solidify at about 32°C)
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4770 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥97.0 %
- Appearance of solution:(10% in ethanol) clear colorless to
- pH:4.0 to 7.0 (30 % at 20°C)
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



