Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:11 °C (closed cup)
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on Anhydrous Substance):Min. 99.8%
- Acidity:Max. 0.0003 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Boiling point:64 - 65 °C
- Density (20/20):0.791 - 0.793
- Evaporation Residue:Max. 5 ppm
- Water:Max. 0.03%
- Absorbance (210 nm):Max. 0.7
- Absorbance (220 nm):Max. 0.3
- Absorbance (230 nm):Max. 0.15
- Absorbance (235 nm):Max. 0.1
- Absorbance (250 nm):Max. 0.02
- Absorbance (260 nm):Max. 0.01
- Transmittance (210 nm):Min. 20%
- Transmittance (220 nm):Min. 50%
- Transmittance (225 nm):Min. 68%
- Transmittance (230 nm):Min. 74%
- Transmittance (235 nm):Min. 80%
- Transmittance (250 nm):Min. 95%
- Transmittance (260 nm):Min. 98%
- Transmittance (260 - 420 nm):Min. 98%
- Fluorescence (as Quinine; 254 nm):Max. 1 ppb
- Conforms Ph. Eur. R 1053200:Passes test
- Conforms Ph. Eur. R1 1053201:Passes test
- Conforms Ph. Eur. R2 1053202:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Number12,5L Aluminium bottle DIN 62 (ZPACK8000412) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 4L Glass bottle DIN 38 (ZPACK13415438)
Packaging Information
SDS_EN_CH_VWRC20864.pdf SDS_EN_DE_VWRC20864.pdf SDS_EN_NL_VWRC20864.pdf SDS_EN_UK_VWRC20864.pdf sds_VWRC20864
SDS
spec_VWRC20864.290 spec_VWRC20864.320 spec_VWRC20864.360 spec_VWRC20864.400 spec_VWRC20864.420
Product specification
More