Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.7 °C (760 mmHg)
Melting Pt:–98 °C
Density:0.791 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS06
:GHS08
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 % (K.F.)
- Boiling point:64 to 65 °C
- Density:(20/4 °C) 0.791 to 0.793
- Solubility:(In water) Passes test
- GC:≥99.8 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Substance darkened by H2SO4:Passes test
- Carbonyl compounds:≤0.001 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0002 meq/g
- KMnO4-reducing substance:Passes test