Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: (Diethylamino)ethane, TEA
CAS Number:121-44-8
:
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
:101.19 g/mol
Merck Index:13,09740
Boiling Pt:90 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
EINECS:204-469-4
UN:1296
ADR:3 (8),II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–11 °C
:GHS02
:GHS07
:GHS05
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.6 %
- IR Spectrum:Passes test
- Refractive index (20°C):1.399 - 1.401
- Evaporation residue:Max. 50 ppm
- Water:Max. 0.2 %
- Fe (Iron):Max. 0.2 ppm
- Pb (Lead):Max. 0.02 ppm
- Transmittance (254 nm):Min. 20 %
- Transmittance (280 nm):Min. 90 %
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Glass bottle for liquids DIN 32 (ZPACK13415030)
Packaging Information
27832921.pdf spec_VWRC28757.184 spec_VWRC28757.184 spec_VWRC28757.230 spec_VWRC28757.230