Order Entry
China
ContactUsLinkComponent
1,2-Dichloroethane 99+% ACS, meets analytical specifications of Reagent Ph. Eur.
Toxic
Health
Flammable
1,2-Dichloroethane 99+% ACS, meets analytical specifications of Reagent Ph. Eur.
Catalog # ACRO406820010
CAS Number:  
undefined
Toxic
Health
Flammable
1,2-Dichloroethane 99+% ACS, meets analytical specifications of Reagent Ph. Eur.
Catalog # ACRO406820010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (Coulometric)
  • Density:
    (20/20°C) about 1.25
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Titrable acid:
    ≤0.0003 meq/g

Specification Test Results

About this item

 :
 :98.96 g/mol
Merck Index:13,03831
Boiling Pt:81…85 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.25 g/cm³
Flash Pt:13 °C
EINECS:203-458-1
UN:1184
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963