Order Entry
China
ContactUsLinkComponent
Supelco® Organic Reference Standard, Stearic acid
Supelco® Organic Reference Standard, Stearic acid
Catalog # SUPL85679-500MG
Supplier:  SUPELCO
CAS Number:  
undefined
Supelco® Organic Reference Standard, Stearic acid
Catalog # SUPL85679-500MG
Supplier:  SUPELCO
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Standards:
    Analytical standard
  • CAS No.:
    57-11-4
  • Description:
    Stearic acid
  • Seq:
    01
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002752
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid
  • Melting point:
    67 - 72 °C (lit.)
  • Formula:
    CH₃(CH₂)₁₆COOH
  • Molecular weight:
    284,48
  • Boiling point:
    361 °C (lit.)

Specifications

About this item