Order Entry
China
ContactUsLinkComponent
Ethyl acetate ≥99.6% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Ethyl acetate ≥99.6% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Ethyl acetate ≥99.6% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-330 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤255
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission, For Information Only:
    For Information Only
  • Maximum for Impurities, For Information Only:
    For Information Only
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission:
    = 99.6 %
  • Assay (by GC, corrected for water):
    ≥99.6 %
  • Residue after Evaporation:
    ≤2.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.8
  • Water (by KF, volumetric):
    ≤0.04 %

Specification Test Results

Related Information