Order Entry
China
ContactUsLinkComponent
 
 
3-Hexanone 98%
 
undefined
 
3-Hexanone 98%
 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3990 to 1.4010 (20°C, 589 nm)
  • GC:
    ≥97.5 %