

Zinc(II) nitrate hexahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10196-18-6
:
Formula:ZnN₂O₆·6H₂O
MDL Number:MFCD00149889
:297.49 g/mol
Merck Index:12,10273
Boiling Pt:∼105 °C (1013 hPa)
Melting Pt:36 °C
Density:2.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-943-8
UN:1514
ADR:5.1,II
:GHS03
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:99.0 - 101.0 %
- IMPORTANT: Store below 24°C (75°F). Ship in refrigerated trucks only.:= 0.005 %
- Assay (Zn(NO₃)₂·6H₂O) (by EDTA titrn):99.0 - 101.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:3.5 - 5.5
- Chloride (Cl):≤0.002 %
- Sulfate (SO₄):≤0.005 %
- Lead (Pb):≤0.005 %
- Trace Impurities - Copper (Cu):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm