


Tetrahydrofuran ≥99.8% contains no preservatives ACS, meets analytical specification of USP for HPLC, for spectrophotometry
HONEAH340-4
:
- Pk:4 L
- Seq:5162
- Color (APHA):20 max.
- GC Analysis:99.8 % min.
- Peroxides (as H₂O₂) at time of packaging:2 mg/L max.
- Refractive Index (20ºC):1.4065 - 1.4075
- Residue:3 mg/L max.
- UV Absorbance @ 250 nm:0.18 AU max.
- UV Absorbance @ 300 nm:0.02 AU max.
- UV Cutoff:212 nm max.
- Water by Karl Fischer Titration:0.03 % max.
:
:72.11 g/mol
:12,09351
:11.8 %(V)
:–108.5 °C
:0.8892 g/cm³ (20 °C)
:–14 °C
:Ambient
:203-726-8
:2056
:3,II
:05-2114615188-48
:GHS02
:GHS08
:GHS07
:109-99-9
:C₄H₈O
:MFCD00005356