Order Entry
China
ContactUsLinkComponent
Sodium metaborate tetrahydrate 98.5%, extra pure
Irritant
Health
Sodium metaborate tetrahydrate 98.5%, extra pure
Catalog # ACRO211635000
CAS Number:  
undefined
Irritant
Health
Sodium metaborate tetrahydrate 98.5%, extra pure
Catalog # ACRO211635000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration with HCl:
    ≥98.0 %

Specification Test Results

About this item

 :
 :137.86 g/mol
Melting Pt:57 °C
EINECS:231-891-6
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:10555-76-7
Formula:BNaO₂·4H₂O
MDL Number:MFCD00149205