


Methanol ≥99.9%, BAKER ANALYZED ULTRA LC/MS™ for LC-MS, tested for UHPLC, J.T.Baker®
BAKR9863-02
: Avantor
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Gradient Elution Test (a.u.) - 254 nm:≤0.01
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (CH₃OH):≥99.9 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Water (H₂O):≤500 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤25.0 ppb
- LC/MS Suitability - Largest Response on ESI- Negative Mode (as 4-Nitrophenol):≤25.0 ppb
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤20 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤5.0 ppb
- Trace Impurities - Copper (Cu):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤5.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤50.0 ppb
- Trace Impurities - Tin (Sn):≤5.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
:
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:11 °C (closed cup)
:Ambient
:200-659-6
:1230
:3,II
:Filtered through a 0.1 micron filter.
:01-2119433307-44
:GHS02
:GHS06
:GHS08
:67-56-1
:CH₃OH
:MFCD00004595