

Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0.782 g/cm³ (20 °C)
EINECS:200-835-2
UN:1648
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:2 °C (closed cup)
REACH:01-2119471307-38
:GHS02
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Evaporation residue:Max. 5 ppm
- Water:Max. 0.02 %
- Absorbance (195 nm):Max. 0.1
- Absorbance (200 nm):Max. 0.05
- Absorbance (210 nm):Max. 0.02
- Absorbance (220 nm):Max. 0.02
- Absorbance (240 nm):Max. 0.005
- Transmittance (195 nm):Min. 80 %
- Transmittance (200 nm):Min. 90 %
- Transmittance (210 nm):Min. 94 %
- Transmittance (220 nm):Min. 96 %
- Transmittance (240 nm):Min. 99 %
- Gradient grade (210 nm):Max. 10 mAU
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC20060.290 spec_VWRC20060.290 spec_VWRC20060.320 spec_VWRC20060.360 spec_VWRC20060.360
Product specification
More