Order Entry
China
ContactUsLinkComponent
Amantadine hydrochloride 99+%
Amantadine hydrochloride 99+%
Certificates
undefined
Amantadine hydrochloride 99+%
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥99.0 %
  • HPLC:
    ≥96.0 %