Order Entry
China
ContactUsLinkComponent
 
 
 
 
Bromoethane 99+%, extra pure
 
undefined
 
 
 
Bromoethane 99+%, extra pure

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4230 to 1.4250 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Acidity:
    ≤0.0005 % (as HBr)