

Tin(II) chloride dihydrate ≥96%, GPR RECTAPUR® for mirror makers
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10025-69-1
:
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863
:225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS07
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:Min. 96 %
- SO4 (Sulphate):Max. 0.02 %
- Fe (Iron):Max. 100 ppm
- Pb (Lead):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC23743.230 spec_VWRC23743.230 spec_VWRC23743.296