Sodium thiosulphate pentahydrate ≥98%, purified
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10102-17-7
:
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186
:248.19 g/mol
Merck Index:12,08844
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 20 ppm
- Fe (Iron):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC27905.293 spec_VWRC27905.293 spec_VWRC27905.362 spec_VWRC27905.362