Order Entry
China
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 99+% for analysis
Sodium dihydrogen phosphate monohydrate 99+% for analysis
Catalog # ACRO207805000
CAS Number:  
undefined
Sodium dihydrogen phosphate monohydrate 99+% for analysis
Catalog # ACRO207805000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • pH:
    4.1 to 4.5 (5 % at 20°C)
  • Titration with NaOH:
    ≥99.0 %
  • Chloride (Cl):
    ≤5 ppm
  • Sulfate (SO4):
    ≤30 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.01 % (Ca and NH4OH ppt)
  • Nitrogen compounds (as N):
    ≤0.001 %

Specification Test Results

About this item

 :
 :137.99 g/mol
Merck Index:13,08734
Density:2,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
CAS Number:10049-21-5
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208