
di-Sodium tetraborate decahydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
:1303-96-4
:
:Na₂B₄O₇·10H₂O
:MFCD00149193
:381.37 g/mol
:13,08662
:320 °C (1013 hPa)
:75 °C
:1.73 g/cm³ (20 °C)
:Ambient
:215-540-4
:01-2119490790-32
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay:99.0 - 103.0 %
- Appearance of solution:Passes test Ph.Eur.
- CO3 (Carbonate):Passes test
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Identification C:Passes test Ph.Eur.
- pH (20°C; 4 %):9.0 - 9.6
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- NH4 (Ammonium):Max. 10 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- As (Arsenic):Max. 2 ppm
- Ca (Calcium):Max. 100 ppm
- Fe (Iron):Max. 5 ppm
- Conforms to Reag. Ph.Eur.:Passes test