Order Entry
China
ContactUsLinkComponent
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # HACH32332
Supplier:  HACH
CAS Number:  
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # HACH32332
Supplier:  HACH
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499