Order Entry
China
ContactUsLinkComponent
Orthophosphoric acid ≥85% in aqueous solution
Corrosive
Orthophosphoric acid ≥85% in aqueous solution
Catalog # ALFAA18067.AP
CAS Number:  
Corrosive
Orthophosphoric acid ≥85% in aqueous solution
Catalog # ALFAA18067.AP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :98 g/mol
Merck Index:14,7344
Melting Pt:19…21 °C
Density:1.700
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340