Order Entry
China
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Sigma-Aldrich®
  SIAL05030-500ML-F
 :  
undefined
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution, Sigma-Aldrich®
  SIAL05030-500ML-F
 :  
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
 :
 :288.38 g/mol
 :1…1.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :Ambient
 :205-788-1
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175