Formic acid 80% ≥80%, TECHNICAL
Supplier: VWR Chemicals
About this item
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:3412
3 Options Available Below
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:3412
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC20315.366_SPEC_EN.pdf P20_VWRC20315.446_SPEC_EN.pdf spec_VWRC20315.297 spec_VWRC20315.297 spec_VWRC20315.366
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications for Product Family
Specification Test Results
- Assay:79 - 81 %
- n 20/D:1.369 - 1.372
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



