Order Entry
China
ContactUsLinkComponent
 
 
 
Chloroform-[D₁] 100.0 atom % D for NMR spectroscopy
  ACRO320680075
 :  
undefined
 
 
Chloroform-[D₁] 100.0 atom % D for NMR spectroscopy
  ACRO320680075
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Pk Info:
    10 x 0.75 ml

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 % (Coulometric)
  • D-Enrichment:
    ≥99.95 %

 

 

 :
 :120.38 g/mol
 :60.8 °C (1013 hPa)
 :–64 °C
 :1.5 g/cm³ (20 °C)
 :Refrigerator
 :212-742-4
 :1888
 :6.1,III
 :GHS06
 :GHS08
 :865-49-6
 :CDCl₃
 :MFCD00000827