Order Entry
China
ContactUsLinkComponent
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Toxic
Health
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Catalog # ACRO217310100
CAS Number:  
undefined
Toxic
Health
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Catalog # ACRO217310100
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    10 mL
  • Seq:
    2076
  • Pk Info:
    10 x 1 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

About this item

 :
 :120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827