Order Entry
China
ContactUsLinkComponent
Hexane (mixture of isomers) 99%, pure
 
 
 
 
Hexane (mixture of isomers) 99%, pure
  ACRO411540010
 :  
undefined
 
 
 
 
Hexane (mixture of isomers) 99%, pure
  ACRO411540010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3770 to 1.3810 (20°C, 589 nm)
  • GC:
    ≥98.5 % (sum of isomers)

 

 

 :
 :86.18 g/mol
 :13,04712
 :68…70 °C (1013 hPa)
 :–95 °C
 :0.659 g/cm³
 :–22 °C
 :Ambient
 :203-777-6
 :1208
 :3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
 :92112-69-1
 :H₃C(CH₂)₄CH₃
 :MFCD00009520