Order Entry
China
ContactUsLinkComponent
Disposable particle filtering respirators, 8247, R95
Disposable particle filtering respirators, 8247, R95
Catalog # MMMA8247
Supplier:  3M
undefined
Disposable particle filtering respirators, 8247, R95
Catalog # MMMA8247
Supplier:  3M
Specifications
  • Type:
    8247
  • Seq:
    01
  • Size:
    U*
  • Environmentally Preferable:
  • Pk:
    20
  • With valve:
    -
  • Class:
    R95
Specifications
About this item
This disposable R95 particulate respirator helps provide reliable respiratory protection against certain oil and non-oil based particles. Constructed with a carbon filter layer for nuisance levels of organic vapors such as solvents, degreasers, and resins. Ideal for a variety of applications, e.g. foundry operations, lab settings, agriculture, petrochemical manufacturing and undercoating.
  • Collapse resistant shell for increased durability
  • Compatible with a variety of protective eyewear and hearing protection
  • Adjustable noseclip reduces potential for eyewear fogging and helps ensure a better seal and fit
  • Two-strap design with dual point attachment helps provide a secure fit
  • Braided headband provides comfort and durability
  • Lightweight construction promotes greater worker comfort and contributes to increased wear time
Recommended for relief against nuisance levels of organic vapors. Nuisance level organic vapor refers to concentrations not exceeding OSHA PEL or applicable exposure limits, whichever is lower.