Bottles, narrow neck, round, low particulate, HDPE, IP2, with screw caps, Nalgene®
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Screw cap supplied assembled on the bottle
- Leakproof
Utilize Nalgene™ Narrow-Mouth HDPE IP2 Bottles as inner containers for designing, assembling, and certifying combination packaging. These high density polyethylene bottles are safe, economical, and designed to resist splitting and puncturing from shipping shocks.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
- Catalog No:
- NALG2099-0001
- NALG2099-0016
- NALG2099-0032
- NALG2099-0008
- NALG2099-0010
- NALG2099-0002
- NALG2099-0004
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)