Tube holder kit (444-7047) consists of:
- Insert retainer
- Tube holder for 10 tubes of Ø 9 mm, 8 tubes of Ø 13 mm
- Tube holder for 8 tubes of Ø 14 mm, 8 tubes of Ø 19 mm
- Tube holder for 8 tubes of Ø 25/29 mm, 5 tubes of Ø 20 mm
- Two vessel harnesses (example, Erlenmeyer)
- Foam insert without holes
Attachment and foam inserts for microplates (444-0210):
Set of two foam inserts for microplates (444-0219) and one insert retainer (444-7048)
Insert holder and foam inserts for microtubes (444-0209):
Set of two foam inserts for microtubes (444-0218) and one insert retainer (444-7048)
Single tube holder (444-0217):
Fits all vortex shakers and holds tubes 64 to 114 mm length, minimum Ø 19 mm. Hands-free mixing. Simple assembly, no tools required. Holds 15 or 50 ml centrifuge tubes.
Ampoule tube adapters (444-0212 and 444-0213)
Clamping devices for up to four vials or tubes. Two sizes available:
For vials/tubes 15 to 17 mm Ø
For vials/tubes 10 to 17 mm Ø
** Can only be used with tube holder 444-0217
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- VWRI444-7058
- VWRI444-7059
- VWRI444-7060
- VWRU12620-874
- VWRI444-0210
- VWRI444-0211
- VWRU12620-898
- VWRI444-0214
- VWRI444-0215
- VWRI444-0212
- VWRI444-0213
- VWRI444-0208
- VWRU58816-156
- VWRU12620-878
- VWRU58816-136
- VWRI444-0209
- VWRU12620-888
- VWRU12620-886
- VWRU58816-154
- VWRI444-7050
- VWRI444-7051
- VWRI444-7052
- VWRI444-7053
- VWRI444-7047
- VWRI444-7048
- VWRU58816-140
- VWRI444-7049
- VWRU58816-130
- VWRU58816-132
- VWRU12620-872
- VWRU12620-896
- VWRU58816-152
- VWRU12620-890
- VWRI444-0218
- VWRI444-0219
- VWRI444-0216
- VWRI444-0217
- VWRU58816-138
- VWRU58816-142
- VWRU12620-876
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing