Stereo microscopes, SZ-51 and SZ-61
Catalog # OLYMSZ51
Supplier: OLYMPUS OPTICAL
Specifications
- Model:SZ51
- Description:Stereo microscope
- Environmentally Preferable:
- Illumination:–
- Magnification:0,8× - 4×
- Pk:1
- Type:Binocular
Specifications
About this item
These stereo microscopes are dedicated for observations in reflected and transmitted light.
- Field of vision close to that achieved through the eyepieces
- Improved observation comfort during long periods
- Good depth of focus
The SZ-51 is equipped with a reflection stage to observe samples up to several centimetres in size.
Ordering information: For your imaging requirements, see camera DP20.