Order Entry
China
ContactUsLinkComponent
Gas-clean filters
Gas-clean filters
Catalog # HEWLCP17973
undefined
Gas-clean filters
Catalog # HEWLCP17973
Specifications
  • Description:
    Carrier gas filter
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Clean gases reduce the risk of column damage and improve accuracy and sensitivity. These unbreakable and self-indicating filters give high sensitivity to oxygen and moisture and can be installed in seconds without tools.