Centrifuge tubes, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PC, round bottom and rimless, autoclavable.
- Transparent
- Withstands forces up to 50000×g
- Suitable for use in refrigerated or non refrigerated centrifuges
Ordering information: 525-2889 and 525-2890 are lipped.
Specifications
- Catalog No:
- NALG3117-9500
- NALG3117-1000
- NALG3117-0500
- NALG3117-0380
- NALG3117-0160
- NALG3117-0150
- NALG3117-0120
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement