Order Entry
China
Orders LinkContactUsLinkComponent
Adapters, distilling heads
Adapters, distilling heads
Supplier:   Lenz Laborglas
Adapters, distilling heads
Supplier:  Lenz Laborglas
Certificates
Adapters, distilling heads
Adapters, distilling heads
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Includes a socket for a distillation thermometer of 55 mm fitted length and two cones at an angle of 75°.
Documents
Product Family Options
Product Information
  • Socket size
    Cone size
    Availability
    Price
  • 14/23 NS
    14/23 - 14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 14/23 NS
    19/26 - 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 14/23 NS
    24/29 - 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 14/23 NS
    24/29 - 24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 14/23 NS
    29/32 - 29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件