Order Entry
China
Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
Supplier:   BOCHEM
Double spatulas, micro
Supplier:  BOCHEM
Certificates
Double spatulas, micro
Double spatulas, micro
8 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 130 mm
    40 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 150 mm
    45 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 150 mm
    45 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 185 mm
    50 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 210 mm
    60 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 210 mm
    60 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 130 mm
    40 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 185 mm
    50 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件