Order Entry
China
Orders LinkContactUsLinkComponent
VWR® Pipette Stands
VWR® Pipette Stands
VWR® Pipette Stands
VWR® Pipette Stands
Pipette stands made of transparent acrylic.
  • Three, four or six-place racks, accommodate most micropipettes
  • Width of upper notches: 25 mm
  • Width of lower notches: 20 mm
Product Family Options
  • Description
    For
    Colour
  • Pipette stand
    3 pipettes
    Transparent
    1 * 1 件
    1 * 1 件 - ¥1,045.00
    ¥1,045.00
    1 * 1 件
       ¥1,045.00
  • Pipette stand
    4 pipettes
    Transparent
    1 * 1 件
    1 * 1 件 - ¥1,112.00
    ¥1,112.00
    1 * 1 件
       ¥1,112.00
  • Pipette stand
    6 pipettes
    Transparent
    1 * 1 件
    1 * 1 件 - ¥1,224.00
    ¥1,224.00
    1 * 1 件
       ¥1,224.00