Graduated pipettes
: BRAND
PP, high clarity.
- Calibrated to deliver (TD, Ex)
- Circular markings and inscriptions in blue
- Cleaning temperature below 60 °C recommended in order to preserve markings and inscriptions
Suction tube outer Ø: 8 mm (except 612-4915: 10 mm)