Bulb pipettes, one mark
Supplier: BRAND
Certificates
About this item
PP, highly translucent.
- Calibrated to deliver (TD, Ex)
- Blue inscriptions
- Exposure to temperatures up to 60 °C will not cause permanent exceeding of tolerance limits
- Cleaning temperature below 60 °C is recommended to preserve marks and inscriptions
- Resistant to breakage