Replication systems, Nunc™
Anodised aluminium replicators with stainless steel pins, PP copiers. Designed for the replication of DNA libraries, this system is ideal for transferring small volumes of liquid from plate to plate or to a membrane. Pins transfer 1,0 µl (±0,1 µl) of liquid.
- Catalog No:
- NUNC250520
- NUNC250393
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement