Tubes, low binding, Nunc-Immuno™, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Graduated polyethylene (PE).
- Surface has lower affinity to proteins than glass and other plastics
- No blocking required
- Compatible with standard equipment
Nunc-Immuno™ MiniSorp™ tubes are suitable for liquid phase immuno-assay methods including RIA.
Specifications
- Catalog No:
- NUNC443990
- NUNC468608
- NUNC466982
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)