Order Entry
China
Orders LinkContactUsLinkComponent
Forceps, teethed
Forceps, teethed
145 mm
Catalog # HAMML-HWC082-14
Supplier:   HAMMACHER KARL
Forceps, teethed
Catalog # HAMML-HWC082-14
Supplier:  HAMMACHER KARL
Forceps, teethed
Forceps, teethed
145 mm
Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 - 12 % nickel)
  • Teethed, straight
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Length:
    145 mm