Multiple vacuum filtration apparatus, Whatman™
Multiple vacuum filtration apparatus suitable only for vacuum operation.
- For microbiological quality control
- Residue analyses
- Serial filtration carried out rapidly and easily with a common drainage outlet
Stainless steel including sieve filter support.
Individual stopcocks, 47/50 mm filter size, tubing nozzle 9 mm vacuum connection.
: Suitable only for vacuum. Do not exceed 1.3bar (300mbar overpressure) when using flushing tubes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.