Dessiccators, vacuum, space saver, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
- Lighter and does not chip like glass
- Holds a full vacuum (760 mm Hg) at room temperature for 24 hours
- White polypropylene bottom models can support incandescent crucibles on Minerit HD high heat ceramic desiccator plates
- Clear polycarbonate bottom models offer increased viewing visibility from more angles
These dessicators feature a tapered polypropylene stopcock with PTFE plug (467-4194) which accepts tubing with 6,35 to 9,525 mm I.D. and allows a controlled entrance of air preventing unnecessary turbulence. Neoprene O-ring assures a tight seal down to 0 °C without the need for grease.
Flat dome, clear polycarbonate top with large ribbed knob maximises interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes.
Ordering information: Supplied with PP plate which is 3,2 mm thick with 3,2 mm diameter holes for air circulation.