Screwthread couplings for threaded pipes, DURAN®
Supplier: DWK LIFE SCIENCES
Certificates
About this item
PBT, red with integrated silicone (VMQ) PTFE seal.
- For flexible connection of two DIN glass threads
- Heat resistant from -45 to +180 °C
- Extremely leak tight