Reversed phase HPLC columns, PRP-3
About this item
PRP-3 polymeric reversed phase columns are ideal for gradient separations of peptides and proteins. The columns are packed with a poly(styrene-divinyl)benzene support.
- Better sample recovery than silica based columns
- Excellent durability
- Last longer than silica based columns
- Crosslinked to prevent shrinkage or swelling
Since the support is entirely polymeric and there are no C18 groups to hydrolyse, column lifetime is longer. Regardless of whether you use ionpair reagents or high percentage aqueous buffers (80 to 100%) for your separation, PRP-3 columns outlast silica based columns under common (pH 2,0) or uncommon (pH 12,7) conditions.
16 Options Available Below
Product Details & Documents
PRP-3 polymeric reversed phase columns are ideal for gradient separations of peptides and proteins. The columns are packed with a poly(styrene-divinyl)benzene support.
- Better sample recovery than silica based columns
- Excellent durability
- Last longer than silica based columns
- Crosslinked to prevent shrinkage or swelling
Since the support is entirely polymeric and there are no C18 groups to hydrolyse, column lifetime is longer. Regardless of whether you use ionpair reagents or high percentage aqueous buffers (80 to 100%) for your separation, PRP-3 columns outlast silica based columns under common (pH 2,0) or uncommon (pH 12,7) conditions.
Specifications for Product Family
Specifications
- Catalog No:
- HAMI79392
- HAMI79466
- HAMI79794
- HAMI79382
- HAMI79574
- HAMI79468
- HAMI79526
- HAMI79147
- HAMI79186
- HAMI79469
- HAMI79461
- HAMI79454
- HAMI79393
- HAMI79395
- HAMI79123
- HAMI79124
- 最大压力:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH 范围:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- 温度限值:
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 5 - 85 °C
- 美国药典代码:
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
Specifications
Product Family Options
Product Information
- 顺序说明列格式Phase硬件栏孔大小颗粒大小内径长度Packing materialAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.HAMI79392最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79466最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79794最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79382最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79574最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79468最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79526最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - Specifications:
More
Specifications
Cat. No.HAMI79147最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - 09Semi-preparativePRP-3Stainless steel300 Å12 - 20 µm21,20 mm100 mmPSDVBLog in for availability1 * 1 件Specifications:
More
Specifications
Cat. No.HAMI79186最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21 - 10Semi-preparativePRP-3Stainless steel300 Å12 - 20 µm21,20 mm250 mmPSDVBLog in for availability1 * 1 件Specifications:
More
Specifications
Cat. No.HAMI79469最大压力5000 psipH 范围1 - 13温度限值5 - 85 °C美国药典代码L21
Analytical guard columns
Product Information
- 顺序说明列格式Phase硬件栏孔大小颗粒大小内径长度Packing materialAvailabilityPrice
- 11Analytical guard column starter kit (1 holder, 2 cartridges)Guard columnPRP-3Stainless steel10 µm2,10 mm20 mmLog in for availability1 * 1 KITSpecifications:
More
Specifications
Cat. No.HAMI79461最大压力pH 范围温度限值美国药典代码 - 12Analytical guard column replacement cartridgesGuard columnPRP-3Stainless steel10 µm2,10 mm20 mmLog in for availability1 * 5 件Specifications:
More
Specifications
Cat. No.HAMI79454最大压力pH 范围温度限值美国药典代码 - 13Analytical guard column starter kit (1 holder, 2 cartridges)Guard columnPRP-3PEEK10 µm3,00 mm8 mmLog in for availability1 * 1 件Specifications:
More
Specifications
Cat. No.HAMI79393最大压力pH 范围温度限值美国药典代码 - 14Analytical guard column replacement cartridgesGuard columnPRP-3PEEK10 µm3,00 mm8 mmLog in for availability1 * 5 件Specifications:
More
Specifications
Cat. No.HAMI79395最大压力pH 范围温度限值美国药典代码
Semi-preparative/preparative guard columns
Product Information
- 顺序说明列格式Phase硬件栏孔大小颗粒大小内径长度Packing materialAvailabilityPrice
- 15Semi-preparative/preparative guard column starter kit (1 holder, 1 cartridge)Guard columnPRP-3Stainless steel12 - 20 µm4,60 mm20 mmLog in for availability1 * 1 件Specifications:
More
Specifications
Cat. No.HAMI79123最大压力pH 范围温度限值美国药典代码 - 16Semi-preparative/preparative guard column replacement cartridgesGuard columnPRP-3Stainless steel12 - 20 µm4,60 mm20 mmLog in for availability1 * 2 件Specifications:
More
Specifications
Cat. No.HAMI79124最大压力pH 范围温度限值美国药典代码



