Order Entry
China
Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
Supplier:   USBECK LABORGERATE
Hoffmann pinch cocks
Supplier:  USBECK LABORGERATE
Certificates
Hoffmann pinch cocks
Hoffmann pinch cocks
10 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
 : The values in the band width column refer to the clearance between the pinch cock legs.
Product Family Options
Standard
Product Information
Heavy duty
Product Information
  • Bandwidth
    Availability
    Price
  • 50 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 60 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 70 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件