Clamps for conical standard ground joints, spring steel, DURAN®
Supplier: DWK LIFE SCIENCES
Certificates
About this item
Stainless steel. Clamps for tight and secure fit of conical ground joint connections.
- Very good chemical resistance, corrosion resistant
- Ideal for use at very high temperatures
- Dimensionally stable, easy to assemble and dismantle