Bottles, wide neck, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polymethylpentene PMP, transparent with PP screw cap.
- High level of chemical resistance
- Temperature resistant up to +145 °C, autoclavable
- Leakproof
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids. Good clarity for viewing bottle contents.
Specifications
- Catalog No:
- NALG2107-0008
- NALG2107-0032
- NALG2107-0016
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement