Dilution bottles, Square, PSF/PPCO, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polysulphone PSF, crystal clear or polypropylene copolymer PPCO, translucent, with PP screw cap.
- PSF: Outstanding resistance to weak acids, bases and all aqueous solutions/PPCO: Outstanding resistance to strong acids, alcohols and bases
- PSF: Temperature resistant from −100 to +165 °C/PPCO: Temperature resistant from −40 to +121 °C
- PSF: Autoclavable, use with Tween shortens service life/PPCO: Autoclavable, sustains repeated cleaning and autoclaving with Tween
Compliant with the dilution methods described in the standard methods for water analysis and the standard methods for the analysis of dairy products as well as other professional standard methods.
Specifications
- Catalog No:
- NALG2505-0380
- NALG2500-0280
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement