Bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Heat resistant up to +100 °C
- Leakproof
Ideal for storing and transporting liquids. Suitable for a wide range of applications.
Specifications
- Catalog No:
- NALG2002-9025
- NALG2002-9050
- NALG2002-0032
- NALG2002-9016
- NALG2002-0016
- NALG2002-0004
- NALG2002-9125
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental certification:
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT Label
NAR Certificate of Retirement - NAR Certificate of Retirement, ACT Label